2-Chlor-3-formyl-6-picolin 97%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-6-methylnicotinaldehyde, 2-Chloro-6-methyl-3-pyridinecarboxaldehyde
OR480236-5G
APOSOR480236-5GEA
136
EUR
APOSOR480236-5G
2-Chlor-3-formyl-6-picolin 97%
2-Chlor-3-formyl-6-picolin
Formel:
C₇H₆ClNO Molecular Weight: 155,5816 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368878 CAS-Nummer: 91591-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...