(S)-(-)-4-Chlor-α-methylbenzylalkohol 95%
Lieferant: Apollo Scientific
Synonyme:
(S)-1-(4-Chlorophenyl)ethanol
OR480238-1G
OR480238-5G
APOSOR480238-1GEA
82.8
EUR
APOSOR480238-1G
APOSOR480238-5G
(S)-(-)-4-Chlor-α-methylbenzylalkohol 95%
(S)-(-)-4-Chlor-α-methylbenzylalkohol
Formel:
C₈H₉ClO Molecular Weight: 156,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797296 CAS-Nummer: 99528-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...