4-(Aminomethyl)pyrimidine hydrochloride 97%
Lieferant: Apollo Scientific
OR480245-1G
OR480245-500MG
OR480245-5G
APOSOR480245-1GEA
303
EUR
APOSOR480245-1G
APOSOR480245-500MG
APOSOR480245-5G
4-(Aminomethyl)pyrimidine hydrochloride 97%
4-(Aminomethyl)pyrimidine hydrochloride
Formel:
C₅H₇N₃·HCl Molecular Weight: 145.59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375466 CAS-Nummer: 1138011-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...