5-Propyl-1H-pyrazol-3-ylamine 97%

Lieferant: Apollo Scientific

Synonyme: 5-Propyl-1H-pyrazol-3-amine

OR480246-10G OR480246-1G OR480246-5G
APOSOR480246-10GEA 628 EUR
APOSOR480246-10G APOSOR480246-1G APOSOR480246-5G
5-Propyl-1H-pyrazol-3-ylamine 97%
5-Propyl-1H-pyrazol-3-ylamine
Formel: C₆H₁₁N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08061095
CAS-Nummer: 126748-58-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR