2,6-Dichloropyrimidine-4-carbonyl chloride 95%
Lieferant: Apollo Scientific
OR480251-10G
OR480251-1G
OR480251-5G
APOSOR480251-10GEA
1160
EUR
APOSOR480251-10G
APOSOR480251-1G
APOSOR480251-5G
2,6-Dichloropyrimidine-4-carbonyl chloride 95%
2,6-Dichloropyrimidine-4-carbonyl chloride
Formel:
C₅HCl₃N₂O Molecular Weight: 211.44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08444236 CAS-Nummer: 26830-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...