(2-Brom-3-pyridinyl)methanol 97%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-3-pyridinemethanol, (2-Bromopyridin-3-yl)methanol, 2-Bromo-3-(hydroxymethyl)pyridine
OR480253-10G
OR480253-1G
OR480253-5G
APOSOR480253-10GEA
651
EUR
APOSOR480253-10G
APOSOR480253-1G
APOSOR480253-5G
(2-Brom-3-pyridinyl)methanol 97%
(2-Brom-3-pyridinyl)methanol
Formel:
C₆H₆BrNO Molecular Weight: 188,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08276300 CAS-Nummer: 131747-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...