(5-Chlor-3-pyridinyl)methanol 97%

Lieferant: Apollo Scientific
Achtung

OR480255-1G OR480255-250MG
APOSOR480255-1GEA 71.2 EUR
APOSOR480255-1G APOSOR480255-250MG
(5-Chlor-3-pyridinyl)methanol 97%
(5-Chlor-3-pyridinyl)methanol
Formel: C₆H₆ClNO
Molecular Weight: 143,5709 g/mol
Schmelzpunkt: 26…28 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09743969
CAS-Nummer: 22620-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR