1-Benzylpyrrolo[2,3-b]pyridine 97%

Lieferant: Apollo Scientific

Synonyme: 1-Benzyl-1H-pyrrolo[2,3-b]pyridine

OR480258-1G OR480258-250MG OR480258-500MG
APOSOR480258-1GEA 1030 EUR
APOSOR480258-1G APOSOR480258-250MG APOSOR480258-500MG
1-Benzylpyrrolo[2,3-b]pyridine 97%
1-Benzylpyrrolo[2,3-b]pyridine
Formel: C₁₄H₁₂N₂
Molecular Weight: 208,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834870
CAS-Nummer: 152955-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR