4-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 95%
Lieferant: Apollo Scientific
OR480259-1G
OR480259-250MG
OR480259-500MG
APOSOR480259-1GEA
524
EUR
APOSOR480259-1G
APOSOR480259-250MG
APOSOR480259-500MG
4-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 95%
4-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Formel:
C₈H₅BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09880126 CAS-Nummer: 1000340-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...