(3-Methylenecyclobutyl)carbamic acid tert-butylester 95%
Lieferant: Apollo Scientific
OR480264-1G
OR480264-500MG
OR480264-5G
APOSOR480264-1GEA
257
EUR
APOSOR480264-1G
APOSOR480264-500MG
APOSOR480264-5G
(3-Methylenecyclobutyl)carbamic acid tert-butylester 95%
(3-Methylenecyclobutyl)carbamic acid tert-butylester
Formel:
C₁₀H₁₇NO₂ Molecular Weight: 183,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09991757 CAS-Nummer: 130369-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...