(2-Oxopyrrolidin-1-yl)phenyl acetic acid, technische Qualität

Lieferant: Apollo Scientific

OR480270-1G OR480270-500MG OR480270-5G
APOSOR480270-1GEA 315 EUR
APOSOR480270-1G APOSOR480270-500MG APOSOR480270-5G
(2-Oxopyrrolidin-1-yl)phenyl acetic acid, technische Qualität
(2-Oxopyrrolidin-1-yl)phenyl acetic acid
Formel: C₁₂H₁₃NO₃
Molecular Weight: 219.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11214950
CAS-Nummer: 60729-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR