1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: 1,4,5,6-Tetrahydro-pyrrolo[3,4-c]pyrazole,hydrochloride

OR480295-1G OR480295-5G
APOSOR480295-1GEA 338 EUR
APOSOR480295-1G APOSOR480295-5G
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride
Formel: C₅H₉Cl₂N₃
Molecular Weight: 182,0511 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08457551
CAS-Nummer: 157327-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR