L-m-Tyrosine 95%

Lieferant: Apollo Scientific

OR480304-1G OR480304-5G
APOSOR480304-1GEA 108 EUR
APOSOR480304-1G APOSOR480304-5G
L-m-Tyrosine 95%
L-m-Tyrosine
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063059
CAS-Nummer: 587-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR