Benzyl-4-chlorphenylketon

Lieferant: Apollo Scientific
Gefahr

Synonyme: Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone

OR4804-100G
APOSOR4804-100GEA 183 EUR
APOSOR4804-100G
Benzyl-4-chlorphenylketon
Benzyl-4-chlorphenylketon
Formel: C₁₄H₁₁ClO
Molecular Weight: 230,69 g/mol
Schmelzpunkt: 103…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016342
CAS-Nummer: 1889-71-0
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR