L-Methionine sulphoximine 98%

Lieferant: Apollo Scientific

Synonyme: (2S)-2-Amino-4-(S-methylsulphonimidoyl)butanoic acid, H-Met(sulphoximine)-OH

OR480433-1G OR480433-5G
APOSOR480433-1GEA 328 EUR
APOSOR480433-1G APOSOR480433-5G
L-Methionine sulphoximine 98%
L-Methionine sulphoximine
Formel: C₅H₁₂N₂O₃S
Molecular Weight: 180,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002621
CAS-Nummer: 15985-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR