2,5-Dichlorobenzothiazole 97%

Lieferant: Apollo Scientific

OR480498-5G
APOSOR480498-5GEA 125 EUR
APOSOR480498-5G
2,5-Dichlorobenzothiazole 97%
2,5-Dichlorobenzothiazole
Formel: C₇H₃Cl₂NS
Molecular Weight: 204,0764 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00044024
CAS-Nummer: 2941-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR