4-Methoxybenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

OR480581-100G OR480581-25G
APOSOR480581-100GEA 259 EUR
APOSOR480581-100G APOSOR480581-25G
4-Methoxybenzolsulfonamid
4-Methoxybenzolsulfonamid
Formel: C₇H₉NO₃S
Molecular Weight: 187,22 g/mol
Schmelzpunkt: 111…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025392
CAS-Nummer: 1129-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR