[(2-Bromethoxy)methyl]benzol 97%

Lieferant: Apollo Scientific

Synonyme: ((2-Bromoethoxy)methyl)benzene, Benzyloxyethyl Bromide, (2-Bromoethoxymethyl)benzene

OR480582-100G
APOSOR480582-100GEA 84.1 EUR
APOSOR480582-100G
[(2-Bromethoxy)methyl]benzol 97%
[(2-Bromethoxy)methyl]benzol
Formel: C₉H₁₁BrO
Molecular Weight: 215,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321307
CAS-Nummer: 1462-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR