4-Ethoxybenzoylchlorid 98%

Lieferant: Apollo Scientific

OR480584-25G
APOSOR480584-25GEA 84.1 EUR
APOSOR480584-25G
4-Ethoxybenzoylchlorid 98%
4-Ethoxybenzoylchlorid
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000688
CAS-Nummer: 16331-46-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR