Boc-N-methyl-D-alanine 98%
Lieferant: Apollo Scientific
Synonyme:
Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine
OR480585-100G
APOSOR480585-100GEA
139
EUR
APOSOR480585-100G
Boc-N-methyl-D-alanine 98%
Boc-N-methyl-D-alanine
Formel:
C₉H₁₇NO₄ Molecular Weight: 203,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037242 CAS-Nummer: 16948-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...