2-Hydroxypyridin-N-oxid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Pyridinol-1-oxide

OR480588-100G OR480588-25G OR480588-5G OR480588-1KG
APOSOR480588-100GEA 49.3 EUR
APOSOR480588-100G APOSOR480588-25G APOSOR480588-5G APOSOR480588-1KG
2-Hydroxypyridin-N-oxid 98%
2-Hydroxypyridin-N-oxid
Formel: C₅H₅NO₂
Molecular Weight: 111,10 g/mol
Schmelzpunkt: 147…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006195
CAS-Nummer: 13161-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR