2-Phenylimidazo[1,2-a]pyridine 95%

Lieferant: Apollo Scientific

OR480608-25G OR480608-5G
APOSOR480608-25GEA 302 EUR
APOSOR480608-25G APOSOR480608-5G
2-Phenylimidazo[1,2-a]pyridine 95%
2-Phenylimidazo[1,2-a]pyridine
Formel: C₁₃H₁₀N₂
Molecular Weight: 194.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00219254
CAS-Nummer: 4105-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR