Ethyl-7-bromo-1H-indole-2-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 7-bromo-1H-indole-2-carboxylate
OR480619-1G
OR480619-5G
OR480619-10G
APOSOR480619-1GEA
21.9
EUR
APOSOR480619-1G
APOSOR480619-5G
APOSOR480619-10G
Ethyl-7-bromo-1H-indole-2-carboxylate 95%
Ethyl-7-bromo-1H-indole-2-carboxylate
Formel:
C₁₁H₁₀BrNO₂ Molecular Weight: 268.1066 g/mol Schmelzpunkt: 87…88 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04972046 CAS-Nummer: 16732-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...