(4-Methoxyphenoxy)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Methoxyphenoxy)acetic acid

OR480633-100G
APOSOR480633-100GEA 266 EUR
APOSOR480633-100G
(4-Methoxyphenoxy)essigsäure 98%
(4-Methoxyphenoxy)essigsäure
Formel: (C₆H₄(OCH₃))OCH₂COOH
Molecular Weight: 182,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00014360
CAS-Nummer: 1877-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR