L-Pipecolsäure 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline
OR480636-100G
OR480636-25G
APOSOR480636-100GEA
265
EUR
APOSOR480636-100G
APOSOR480636-25G
L-Pipecolsäure 97%
L-Pipecolsäure
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005981 CAS-Nummer: 3105-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...