2-Amino-5-methoxybenzonitril 94%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5-methoxy-benzonitrile

OR480639-5G
APOSOR480639-5GEA 115 EUR
APOSOR480639-5G
2-Amino-5-methoxybenzonitril 94%
2-Amino-5-methoxybenzonitril
Formel: C₈H₈N₂O
Molecular Weight: 148,1600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05149280
CAS-Nummer: 23842-82-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR