2-Iod-5-nitrotoluol 97%
Lieferant: Apollo Scientific
Synonyme:
1-Iodo-2-methyl-4-nitrobenzene
OR480645-100G
APOSOR480645-100GEA
142
EUR
APOSOR480645-100G
2-Iod-5-nitrotoluol 97%
2-Iod-5-nitrotoluol
Formel:
C₇H₆INO₂ Molecular Weight: 263,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051517 CAS-Nummer: 5326-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...