2,2,6,6-Tetrachlorcyclohexanon 95%

Lieferant: Apollo Scientific

OR480659-25G OR480659-5G
APOSOR480659-25GEA 372 EUR
APOSOR480659-25G APOSOR480659-5G
2,2,6,6-Tetrachlorcyclohexanon 95%
2,2,6,6-Tetrachlorcyclohexanon
Formel: C₆H₆Cl₄O
Molecular Weight: 235,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00019451
CAS-Nummer: 3776-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR