3-Phenylpyridin 95%

Lieferant: Apollo Scientific

OR480660-100G OR480660-25G
APOSOR480660-100GEA 525 EUR
APOSOR480660-100G APOSOR480660-25G
3-Phenylpyridin 95%
3-Phenylpyridine
Formel: C₁₁H₉N
Molecular Weight: 155,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006380
CAS-Nummer: 1008-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR