2,5-Dimethoxyphenylisocyanat 98%
Lieferant: Apollo Scientific
Synonyme: 2-Isocyanato-1,4-dimethoxybenzene
OR480661-100G OR480661-25G
APOSOR480661-100GEA 563 EUR
APOSOR480661-100G APOSOR480661-25G
2,5-Dimethoxyphenylisocyanat 98%
2,5-Dimethoxyphenylisocyanat
  • May form small amount of sediment.
Formel: C₉H₉NO₃
Molecular Weight: 179.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002006
CAS-Nummer: 56309-62-7