N-ɑ-Boc-N(in)-formyl-L-tryptophan 97+%

Lieferant: Apollo Scientific

Synonyme: N-Boc-1-formyl-L-tryptophan, N-ɑ-(tert-Butoxycarbonyl)-N1-formyl-L-tryptophan, N-ɑ-Boc-N1-formyl-L-tryptophan, Boc-Trp(For)-OH, Boc-Trp(CHO)-OH

OR480669-100G
APOSOR480669-100GEA 343 EUR
APOSOR480669-100G
N-ɑ-Boc-N(in)-formyl-L-tryptophan 97+%
N-alpha-Boc-N(in)-formyl-L-tryptophan
Formel: C₁₇H₂₀N₂O₅
Molecular Weight: 332,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00065992
CAS-Nummer: 47355-10-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR