2-Nitro-4-thiocyanatoanilin 95%

Lieferant: Apollo Scientific

Synonyme: (4-Amino-3-nitro-phenyl) thiocyanate

OR480684-100G
APOSOR480684-100GEA 177 EUR
APOSOR480684-100G
2-Nitro-4-thiocyanatoanilin 95%
2-Nitro-4-thiocyanatoanilin
Formel: C₇H₅N₃O₂S
Molecular Weight: 195,2000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463878
CAS-Nummer: 54029-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR