(2-Brom-4-methoxyphenyl)essigsäure 97%
Lieferant: Apollo Scientific
Synonyme:
2-(2-Bromo-4-methoxyphenyl)acetic acid, 2-Bromo-4-methoxyphenylacetic acid
OR480685-25G
OR480685-5G
APOSOR480685-25GEA
232
EUR
APOSOR480685-25G
APOSOR480685-5G
(2-Brom-4-methoxyphenyl)essigsäure 97%
(2-Brom-4-methoxyphenyl)essigsäure
Formel:
C₉H₉BrO₃ Molecular Weight: 245,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02664700 CAS-Nummer: 66916-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...