Isoquinoline-7-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR48070-1G OR48070-250MG
APOSOR48070-1GEA 122 EUR
APOSOR48070-1G APOSOR48070-250MG
Isoquinoline-7-carbonitrile
Isoquinoline-7-carbonitrile
Formel: C₁₀H₆N₂
Molecular Weight: 154.1680 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699231
CAS-Nummer: 223671-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR