(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%

Lieferant: Apollo Scientific

Synonyme: N-Boc-L-serine benzyl ester, Boc-Ser-OBzl, BOC-L-Serine benzyl ester

OR480702-25G OR480702-100G
APOSOR480702-25GEA 87 EUR
APOSOR480702-25G APOSOR480702-100G
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate
Formel: C₁₅H₂₁NO₅
Molecular Weight: 295,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00076983
CAS-Nummer: 59524-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR