4-Bromresorcin 95%
Lieferant: Apollo Scientific
Synonyme:
4-Bromresorcinol
OR480711-10G
OR480711-25G
OR480711-5G
APOSOR480711-10GEA
94.4
EUR
APOSOR480711-10G
APOSOR480711-25G
APOSOR480711-5G
4-Bromresorcin 95%
4-Bromresorcin
Formel:
C₆H₅BrO₂ Molecular Weight: 189,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002272 CAS-Nummer: 6626-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...