5-Brom-1H-indol-3-carbonsäure 94%
Lieferant: Apollo Scientific
Synonyme:
5-Bromoindole-3-carboxylic acid
OR480712-25G
APOSOR480712-25GEA
200
EUR
APOSOR480712-25G
5-Brom-1H-indol-3-carbonsäure 94%
5-Bromo-1H-indole-3-carboxylic acid
Formel:
C₉H₆BrNO₂ Molecular Weight: 240,06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664007 CAS-Nummer: 10406-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...