2-Amino-6-chloro-4-(diethylamino)pyrimidine 97%

Lieferant: Apollo Scientific

OR480719-10G OR480719-5G
APOSOR480719-10GEA 306 EUR
APOSOR480719-10G APOSOR480719-5G
2-Amino-6-chloro-4-(diethylamino)pyrimidine 97%
2-Amino-6-chloro-4-(diethylamino)pyrimidine
Formel: C₈H₁₃ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00664696
CAS-Nummer: 116895-09-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR