6-Chlor-2,3-pyridindiamin 97%

Lieferant: Apollo Scientific

Synonyme: 2,3-Diamino-6-chloropyridine

OR480720-25G OR480720-5G
APOSOR480720-25GEA 184 EUR
APOSOR480720-25G APOSOR480720-5G
6-Chlor-2,3-pyridindiamin 97%
6-Chloropyridine-2,3-diamine
Formel: C₅H₆ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00209966
CAS-Nummer: 40851-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR