2-Amino-5-phenylpyridin 97%
Lieferant: Apollo Scientific
Synonyme:
5-Phenyl-2-pyridylamin, 5-Phenylpyridin-2-amin
OR480727-5G
APOSOR480727-5GEA
71.2
EUR
APOSOR480727-5G
2-Amino-5-phenylpyridin 97%
2-Amino-5-phenylpyridine
Formel:
C₁₁H₁₀N₂ Molecular Weight: 170,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01692452 CAS-Nummer: 33421-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...