3-Bromo-4-iodopyridine 95%

Lieferant: Apollo Scientific

OR480764-1G OR480764-5G
APOSOR480764-1GEA 82.8 EUR
APOSOR480764-1G APOSOR480764-5G
3-Bromo-4-iodopyridine 95%
3-Bromo-4-iodopyridine
Formel: C₅H₃BrIN
Lagertemperatur: Raumtemperatur
MDL: MFCD11110263
CAS-Nummer: 89167-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR