4-Amino-6-bromopyrimidine 98%
Lieferant: Apollo Scientific
Synonyme:
6-Bromopyrimidin-4-amine
OR480783-5G
APOSOR480783-5GEA
63.8
EUR
APOSOR480783-5G
4-Amino-6-bromopyrimidine 98%
4-Amino-6-bromopyrimidine
Formel:
C₄H₄BrN₃ Molecular Weight: 173,9987 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233963 CAS-Nummer: 1159818-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...