3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%

Lieferant: Apollo Scientific

Synonyme: 1-(4-Chloro-phenyl)-1H-pyrazol-3-ylamine

OR480789-1G OR480789-5G
APOSOR480789-1GEA 339 EUR
APOSOR480789-1G APOSOR480789-5G
3-Amino-1-(4-chlorophenyl)-1H-pyrazole 95%
3-Amino-1-(4-chlorophenyl)-1H-pyrazole
Formel: C₉H₈ClN₃
Molecular Weight: 193,64 g/mol
Schmelzpunkt: 107…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027932
CAS-Nummer: 66000-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR