3-Brompyridazin 95%

Lieferant: Apollo Scientific

OR480809-1G OR480809-5G
APOSOR480809-1GEA 74.1 EUR
APOSOR480809-1G APOSOR480809-5G
3-Brompyridazin 95%
3-Bromopyridazine
Formel: C₄H₃BrN₂
Molecular Weight: 158.9840 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10688587
CAS-Nummer: 88491-61-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR