1H-imidazole-4-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Imidazole-4-sulphonyl chloride

OR480821-10G OR480821-1G OR480821-5G
APOSOR480821-10GEA 687 EUR
APOSOR480821-10G APOSOR480821-1G APOSOR480821-5G
1H-imidazole-4-sulphonyl chloride 95%
1H-imidazole-4-sulphonyl chloride
Formel: C₃H₃ClN₂O₂S
Molecular Weight: 166,579 g/mol
Schmelzpunkt: 163…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08687938
CAS-Nummer: 58767-51-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR