2-Methoxybenzenesulphonyl chloride 95%

Lieferant: Apollo Scientific

OR480823-5G
APOSOR480823-5GEA 79.9 EUR
APOSOR480823-5G
2-Methoxybenzenesulphonyl chloride 95%
2-Methoxybenzenesulphonyl chloride
Formel: C₇H₇ClO₃S
Molecular Weight: 206,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01961367
CAS-Nummer: 10130-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR