2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-chloro-5-methylthiazole-4-carboxylate

OR480827-5G
APOSOR480827-5GEA 169 EUR
APOSOR480827-5G
2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester 95%
2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester
Formel: C₇H₈ClNO₂S
Molecular Weight: 205.66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443895
CAS-Nummer: 907545-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR