Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1-(hydroxymethyl)cyclopropanecarboxylate

OR480832-1G OR480832-5G
APOSOR480832-1GEA 105 EUR
APOSOR480832-1G APOSOR480832-5G
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate 95%
Ethyl-1-(hydroxymethyl)cyclopropanecarboxylate
Formel: C₇H₁₂O₃
Molecular Weight: 144,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09759206
CAS-Nummer: 3697-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR