2-Benzoylcyclohexanecarboxylic acid 95%

Lieferant: Apollo Scientific

OR480857-1G OR480857-5G
APOSOR480857-1GEA 226 EUR
APOSOR480857-1G APOSOR480857-5G
2-Benzoylcyclohexanecarboxylic acid 95%
2-Benzoylcyclohexanecarboxylic acid
Formel: C₁₄H₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07775665
CAS-Nummer: 6939-99-7
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR