6-Isochinolincarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Isoquinolinecarboxylic Acid
OR48100-1G
OR48100-250MG
OR48100-5G
APOSOR48100-1GEA
108
EUR
APOSOR48100-1G
APOSOR48100-250MG
APOSOR48100-5G
6-Isochinolincarbonsäure
6-Isoquinolinecarboxylic acid
Formel:
C₁₀H₇NO₂ Molecular Weight: 173.1681 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09910323 CAS-Nummer: 106778-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...